Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetamide oxime, 95+%, Acros Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
22059-22-9
C2H6N2O
74.08
MFCD00506021
AEXITZJSLGALNH-UHFFFAOYSA-N
n'-hydroxyacetimidamide,n-hydroxyacetamidine,methylamidoxime,acetamide, oxime,1z-n'-hydroxyethanimidamide,n-hydroxyethanimidamide,oximinoacetamide,e-n'-hydroxyacetimidamide,n'-hydroxyacetamidine,n-hydroxy-acetamidine
5487681
N'-hydroxyethanimidamide
CC(=NO)N
22059-22-9
C2H6N2O
74.08
MFCD00506021
AEXITZJSLGALNH-UHFFFAOYSA-N
n'-hydroxyacetimidamide,n-hydroxyacetamidine,methylamidoxime,acetamide, oxime,1z-n'-hydroxyethanimidamide,n-hydroxyethanimidamide,oximinoacetamide,e-n'-hydroxyacetimidamide,n'-hydroxyacetamidine,n-hydroxy-acetamidine
5487681
N'-hydroxyethanimidamide
CC(=NO)N
Color White
Melting Point 134.0°C to 138.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95+%
Linear Formula CH3C(NH2)=NOH
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,insoluble in ether and pentane
Formula Weight 74.08
Physical Form Powder, Crystals and/or Chunks
Percent Purity ≥95%
Chemical Name or Material Acetamide oxime
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.