Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Acetamidocinnamic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
5469-45-4
C11H11NO3
205.21
XODAOBAZOQSFDS-YFHOEESVSA-N
z-2-acetamido-3-phenylacrylic acid,alpha-acetamidocinnamic acid,.alpha.-acetamidocinnamic acid,.alpha.-acetaminocinnamic acid,acetamidocinnamic acid,z-2-acetamido-3-phenylprop-2-enoic acid,2-acetylamino-3-phenyl-2-propenoic acid,2-acetamido-3-phenylprop-2-enoic acid,2z-2-acetamido-3-phenylprop-2-enoic acid,pubchem8139
5370579
(Z)-2-acetamido-3-phenylprop-2-enoic acid
CC(=O)NC(=CC1=CC=CC=C1)C(=O)O
5469-45-4
C11H11NO3
205.21
XODAOBAZOQSFDS-YFHOEESVSA-N
z-2-acetamido-3-phenylacrylic acid,alpha-acetamidocinnamic acid,.alpha.-acetamidocinnamic acid,.alpha.-acetaminocinnamic acid,acetamidocinnamic acid,z-2-acetamido-3-phenylprop-2-enoic acid,2-acetylamino-3-phenyl-2-propenoic acid,2-acetamido-3-phenylprop-2-enoic acid,2z-2-acetamido-3-phenylprop-2-enoic acid,pubchem8139
5370579
(Z)-2-acetamido-3-phenylprop-2-enoic acid
CC(=O)NC(=CC1=CC=CC=C1)C(=O)O
Packaging Glass bottle
Melting Point 188.0°C to 190.0°C
Quantity 100g
Assay Percent Range 97.5% min. (HPLC)
Linear Formula C6H5CH=C(NHCOCH3)CO2H
Beilstein 10,683
Formula Weight 205.21
Percent Purity 98%
Chemical Name or Material α-Acetamidocinnamic acid

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath

GHS Signal Word: Warning

EINECSNumber : 226-795-6