Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetaminophen, Granular, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
103-90-2
C8H9NO2
151.17
RZVAJINKPMORJF-UHFFFAOYSA-N
N-(4-hydroxyphenyl)acetamide
CC(=O)NC1=CC=C(O)C=C1
103-90-2
C8H9NO2
151.17
RZVAJINKPMORJF-UHFFFAOYSA-N
N-(4-hydroxyphenyl)acetamide
CC(=O)NC1=CC=C(O)C=C1

A1278, 103-90-2, C8H9NO2

Spectrum™ Acetaminophen, Granular, USP , also known as paracetamol, is a widely used antipyretic and mild analgesic that is available over-the-counter. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Pail
Ignition Residue 0.001
Quantity 2.5 kg
Loss on Drying 0.005
CAS Max % 1
Percent Purity 98 to 102%