Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetaminophen, Granular, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
103-90-2
C8H9NO2
151.17
RZVAJINKPMORJF-UHFFFAOYSA-N
N-(4-hydroxyphenyl)acetamide
CC(=O)NC1=CC=C(O)C=C1
103-90-2
C8H9NO2
151.17
RZVAJINKPMORJF-UHFFFAOYSA-N
N-(4-hydroxyphenyl)acetamide
CC(=O)NC1=CC=C(O)C=C1

A1278, 103-90-2, C8H9NO2

Spectrum™ Acetaminophen, Granular, USP , also known as paracetamol, is a widely used antipyretic and mild analgesic that is available over-the-counter. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Loss on Drying 0.5%
Ignition Residue 0.1%
Packaging Poly Pail
Quantity 12 kg
CAS Max % 100%
Percent Purity 98 to 102%