Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Acetate, 1M buffer soln., pH, 3.0, low endotoxin

Click to view available options
Quantity:
100mL
250mL
127-09-3
C2H3NaO2
82.034
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
517045
CHEBI:32954
sodium;acetate
CC(=O)[O-].[Na+]
127-09-3
C2H3NaO2
82.034
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
517045
CHEBI:32954
sodium;acetate
CC(=O)[O-].[Na+]
Concentration 1M buffer soln.
Physical Form Liquid
pH 3.0
Color Colorless
Quantity 250mL
Chemical Name or Material Acetate, low endotoxin