Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Acetate Buffer (1 M, pH 4.0)

Catalog No. p-7041749
Click to view available options
Quantity:
250mL
500mL

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol

Sodium Acetate is used as a buffer in the buffering range of pH 3.6-5.6. Other reported uses. Used in the purification and precipitation of nucleic acids; Used in protein crystallization. Staining of gels in protein gel electrophoresis and HPLC; Mordant dyeing. DNA microarray studies of E. coli response; Investigation of protein unfolding during reverse phase chromatography. Used in conjunction with sodium carbonate to enhance the activation of freeze-dried subtilisin Carlsberg in organic solvents and as a substrate of acetokinase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium Acetate is used as a buffer in the buffering range of pH 3.6-5.6. Other reported uses. Used in the purification and precipitation of nucleic acids; Used in protein crystallization. Staining of gels in protein gel electrophoresis and HPLC; Mordant dyeing. DNA microarray studies of E. coli response; Investigation of protein unfolding during reverse phase chromatography. Used in conjunction with sodium carbonate to enhance the activation of freeze-dried subtilisin Carlsberg in organic solvents and as a substrate of acetokinase.

Solubility
Fully miscible with water. Soluble in ethanol.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-09-3

Specifications

Concentration 1M buffer soln.
Molecular Formula C2H3NaO2
MDL Number MFCD00012459
pH Range 4
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
SMILES [Na+].CC([O-])=O
IUPAC Name sodium acetate
Molecular Weight (g/mol) 82.03
Form Liquid
Quantity 250mL
Storage Requirements Ambient temperatures
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.