Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetate Buffer, for Iron (Phenanthroline Method), Ricca Chemical
For Analysis of Iron (pHenanthroline Method)
$105.80 - $906.50
Specifications
Chemical Name or Material | Acetate Buffer |
---|---|
CAS | 64-19-7, 631-61-8, 7732-18-5 |
CAS Min % | 63.0, 21.6, 12.95 |
CAS Max % | 66.0, 22.48, 13.48 |
Color | Colorless |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
6016
|
Ricca Chemical Company
6016 |
500 mL | Natural Poly Bottle |
Each for $105.80
|
|
||||
60-32
|
Ricca Chemical Company
6032 |
1 L | Natural Poly Bottle |
Each for $182.60
|
|
||||
601
|
Ricca Chemical Company
601 |
4 L | Natural Poly Bottle |
Each for $170.10
|
|
||||
R006000020F
|
Ricca Chemical Company
605 |
20 L | Cubitainer |
Each for $906.50
|
|
||||
Specifications
Acetate Buffer | |
63.0, 21.6, 12.95 | |
Colorless |
64-19-7, 631-61-8, 7732-18-5 | |
66.0, 22.48, 13.48 | |
Liquid |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title