Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetate Buffer, pH 3.5 For USP Heavy Metals Test, Reagents
Click to view available options
Quantity:
120 mL
250 mL
500 mL
1 L
4 L
20 L
Specifications
Specifications
Chemical Name or Material | Acetate Buffer, pH 3.5 For USP Heavy Metals Test |
CAS | 7732-18-5, 631-61-8, 631-61-8 |
Boiling Point | 118°C |
Color | Colorless |
pH | 3.5 |
Physical Form | Liquid |
Quantity | 20 L |
Solution Type | Buffer |
Packaging | Cubitainer |
Specific Gravity | 1.0737 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction