Acetate Buffer Solution, pH 3.5, For USP Heavy Metals Testing, Ricca Chemical™

$203.85

Chemical Identifiers

Chemical Name or Material Acetate Buffer Solution
CAS [7732-18-5, 63.00-66.00%], [7647-01-0, 18.71-19.47%], [631-61-8, 15.87-16.51%]
Physical Form Clear, colorless liquid
View More Specs

  • Products 1
  • Safety and Handling
Products
Catalog Number Mfr. No. Description Price Quantity Availability  

60132

Ricca Chemical
60132
1L; Poly natural
Each for $203.85
Check Availability
Add to Cart
Descriptions & Specifications
Safety and Handling

Related Products