Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Fluka™ Acetate Buffer Solution Ph 4.6, Honeywell Fluka™

Click to view available options
Quantity:
500mL
1L
126-96-5
C4H8NaO4
143.094
MFCD00137248
OLFJVIXNILIZKF-UHFFFAOYSA-N
87315833
acetic acid;sodium
CC(=O)O.CC(=O)O.[Na]
126-96-5
C4H8NaO4
143.094
MFCD00137248
OLFJVIXNILIZKF-UHFFFAOYSA-N
87315833
acetic acid;sodium
CC(=O)O.CC(=O)O.[Na]

Sodium acetate / acetic acid, for complexometry

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
UN Number NONH for all modes of transport
Beilstein 3786729
Quantity 500mL
Chemical Name or Material Acetate Buffer Solution Ph 4.6