Learn More
Acetate Buffer TS, (U.S.P. Test Solution), Spectrum™
A-281
Manufacturer:  Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner A2811LT
 View more versions of this product
Catalog No. 18-600-195
Add to cart
Description
Description
Spectrum™ Acetate Buffer TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
Specifications
Specifications
64-19-7, 7732-18-5
Poly Bottle
1 L
0.1%, 99.9%
5.9-6.0