Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetazolamide, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
59-66-5
C4H6N4O3S2
222.24
BZKPWHYZMXOIDC-UHFFFAOYSA-N
N-(5-sulfamoyl-1,3,4-thiadiazol-2-yl)acetamide
CC(=O)NC1=NN=C(S1)S(N)(=O)=O
59-66-5
C4H6N4O3S2
222.24
BZKPWHYZMXOIDC-UHFFFAOYSA-N
N-(5-sulfamoyl-1,3,4-thiadiazol-2-yl)acetamide
CC(=O)NC1=NN=C(S1)S(N)(=O)=O

A1266, 59-66-5, C4H6N4O3S2

Spectrum™ Acetazolamide, USP is in the diuretic and carbonic anhydrase inhibitor families of medications. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Quantity 25 g
Ignition Residue 0.001
CAS Max % 1
Percent Purity 98 to 102%