Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetic acid, 1.0N Standardized Solution, Alfa Aesar™
Click to view available options
Quantity:
1L
4L
20L
Specifications
Appearance | Liquid |
Solubility Information | Miscible with water. |
Formula Weight | 60.05 |
Physical Form | Liquid |
Quantity | 20L |
Chemical Name or Material | Acetic acid |
Safety and Handling
EINECSNumber : 200-580-7
TSCA : Yes
Recommended Storage : Ambient temperatures