Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid, 20% v/v, Reagents

Catalog No. 01335908
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Catalog No. Quantity
01-335-908 20 L
01-335-910 10 L
01-335-909 1 L
01-335-907 4 L
01-335-906 500 mL
5 options

Catalog No. 01-335-908

Supplier: Reagents Holdings Llc CA20420020F

Only null left
Add to Cart
Add to Cart

Acetic Acid, 20% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 64-19-7
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Ethanoic Acid, Ethylic Acid
IUPAC Name water
SMILES O

Specifications

pH 2
Boiling Point 118°C
Color Colorless
Percent Purity 20%
Grade Reagent
Quantity 20 L
Packaging Cubitainer
Physical Form Liquid
Specific Gravity 1.0334
Chemical Name or Material Acetic Acid, 20% v/v
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.