Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid, 50% (w/v) Aqueous Solution, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Cubitainer
Poly Bottle
Poly Drum
64-19-7,7732-18-5
64-19-7,7732-18-5
Percent Purity 50% ± 0.6%
Grade Chemical Solution
Packaging Poly Bottle
Quantity 4 L

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.