Acetic Acid, 80% w/w (Certified), Fisher Chemical

Packaging Glass Bottle
Quantity 4L

 View more versions of this product

Catalog No. A37-4

  Check Availability
Add to Cart

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 64-19-7,7732-18-5
    Assay Percent Range 80 to 82 %
    Molecular Formula C2H4O2
    Linear Formula CH3COOH
    Formula Weight 60.05
    MDL Number MFCD00036152
    Chemical Name or Material Acetic Acid, 80% (w/w)
    Assay 80 to 82%
    Grade Certified Grade

    Specifications

    Melting Point −7°C
    Boiling Point 117°C
    Formula Weight 60.05
    Quantity 4L
    DOT Information DOT Class 8, : Corrosive
    pH 2.4
    Color Colorless
    Physical Form Liquid
    Normality 14.2
    Vapor Pressure 16mmHg at 25°C
    Health Hazard 3 Emergency Overview
    Combustible liquid. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

    NFPA

    Health:3
    Flammability:2
    Instability:0
    Substances Reducing Permanganate Pass Test
    Sulfate 4ppm max.
    Assay Percent Range 80 to 82 %
    Health Hazard 2 DANGER!
    Iron (Fe) 4ppm max.
    Packaging Glass Bottle
    Residue after Evaporation 0.002% max.
    Cleanup Symbol Acid
    Chloride 4ppm max.
    MDL Number MFCD00036152
    ChemAlert Storage Symbol White
    Heavy Metals (as Pb) 4ppm max.
    Identification Pass Test
    CAS 7732-18-5,64-19-7
    Molecular Formula C2H4O2
    Linear Formula CH3COOH
    Formic or Sulfurous Acid Pass Test
    Additional Information Vapor Pressure: 16mmHg at 25°C
    Chemical Name or Material Acetic Acid, 80% (w/w)
    Assay 80 to 82%
    Grade Certified Grade
       

    DANGER!

    Emergency Overview
    Combustible liquid. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

    NFPA

    Health:3
    Flammability:2
    Instability:0

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit