Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic acid, 99.5%, pure, Thermo Scientific Chemicals

Catalog Number AC124040050
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Glass Bottle
Plastic drum
64-19-7
C2H4O2
60.05
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
176
CHEBI:15366
acetic acid
CC(O)=O
64-19-7
C2H4O2
60.05
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
176
CHEBI:15366
acetic acid
CC(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 16.0°C to 16.5°C
Boiling Point 117.0°C to 118.0°C
CAS Min % 99.4
CAS Max % 100.0
Assay Percent Range 99.5%
Linear Formula CH3COOH
Beilstein 02, 96
Fieser 02,5; 05,3; 07,1; 08,1; 12,568; 13,45; 15,12
Merck Index 15, 54
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol, glycerol, ether and, carbon tetrachloride, practically insoluble in carbon disulfide
Formula Weight 60.05
Percent Purity 99.5%
Grade Pure
Density 1.0480g/mL
Quantity 5 L
Flash Point 40°C
Infrared Spectrum Authentic
Packaging Plastic drum
Physical Form Liquid
Refractive Index 1.3700 to 1.3730
Specific Gravity 1.048
Chemical Name or Material Acetic acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit