Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic acid, 99.8%, for biochemistry, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
10L
Packaging:
Glass Bottle
Plastic drum
64-19-7
C2H4O2
60.052
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,acetasol,acide acetique,essigsaeure
176
CHEBI:15366
acetic acid
CC(=O)O
64-19-7
C2H4O2
60.052
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,acetasol,acide acetique,essigsaeure
176
CHEBI:15366
acetic acid
CC(=O)O
Absorbance (neat),(neat) at 0nm,0.02 max. at 280nm,0.1 max. at 260nm
Absorbance Conditions (neat)
CAS Min % 99.75
CAS Max % 100.0
Additional Information Vapor Pressure: 1.52kPa at 20°C
Cadmium (Cd) 0.1ppm max.
Assay Percent Range 99.8%
Linear Formula CH3COOH
Chloride 1ppm max.
Beilstein 02, 96
Fieser 02,5; 05,3; 07,1; 08,1; 12,568; 13,45; 15,12
Merck Index 15, 54
Density 1.0480g/mL
Dilution Test Passes Test
Formula Weight 60.05
Physical Form Liquid
Percent Purity 99.8%
Grade Biochemical
Heavy Metals (as Pb) 5ppm max.
Infrared Spectrum Authentic
Iron (Fe) 0.2ppm max.
Nickel (Ni) 0.1ppm max.
Packaging Plastic drum
Refractive Index 1.3700 to 1.3730
Specific Gravity 1.048
Solubility Solubility in water: miscible. Other solubilities: miscible with alcohol, glycerol, ether and, carbon tetrachloride, practically insoluble in carbon disulfide
Substances Reducing Permanganate Passes Test
Substit Aromatic Aldehydes Passes Test
Substances Reducing Dichromate Passes Test
Sulfate 1ppm max.
Melting Point 16.0°C to 16.5°C
Color Undesignated
Boiling Point 117.0°C to 118.0°C
Quantity 10L
Flash Point 40°C
Chemical Name or Material Acetic acid, For biochemistry
Show More Show Less

GHS P Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Danger