Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetic Acid, Certified, 10.0% (v/v) ±0.5%, LabChem™
Click to view available options
Quantity:
500 mL
1 L
4 L
Specifications
Color | Colorless |
CAS Max % | 89.66, 10.34 |
Linear Formula | CH3COOH |
UN Number | UN2790 |
Solubility Information | Soluble in water |
Formula Weight | 60.05 |
Percent Purity | 10.0% ± 0.5% (v/v) |
Density | 1.01g/mL |
Grade | Certified |
Quantity | 500 mL |
Show More |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. |
Precautionary Statement | GHS P Statement Do not breathe mist, vapors, spray. Wear protective gloves/eye protection. Wash exposed skin thoroughly after handling. If swallowed: Rinse mouth. Do not induce vomiting. Immediately call a poison center/doctor. If on skin (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. Immediately call a poison center/doctor. Wash contaminated clothing before reuse. If inhaled: Remove person to fresh air and keep comfortable for breathing. Immediately call a poison center/doctor. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up. Dispose of contents/container to comply with local, state and federal regulations. |
Recommended Storage | Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction