Alfa Aesar™ Acetic acid, Environmental Grade, 99% min

Manufacturer: Alfa Aesar™ 38740K4

Quantity 2.5L

 View more versions of this product

Catalog No. AA38740K4

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 64-19-7
    Formula Weight 60.05
    MDL Number MFCD00036152
    UN Number UN2789
    Merck Index 14,55
    Chemical Name or Material Acetic acid
    Grade Environmental Grade

    Specifications

    Formula Weight 60.05
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 2.5L
    Density 1.049g/mL
    Odor Pungent
    Refractive Index 1.3721
    TSCA TSCA
    Melting Point 16.6°C
    pH 2.5
    Flash Point 40° (104°F)
    Physical Form Liquid
    MDL Number MFCD00036152
    Health Hazard 1 Danger
    Boiling Point 118.1°C
    CAS 64-19-7
    UN Number UN2789
    Merck Index 14,55
    Health Hazard 2 GHS H Statement
    H314-H226
    Causes severe skin burns and eye damage.
    Flammable liquid and vapor.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ACETIC ACID, GLACIAL
    Chemical Name or Material Acetic acid
    Assay ≤99%
    Grade Environmental Grade

    GHS H Statement
    H314-H226
    Causes severe skin burns and eye damage.
    Flammable liquid and vapor.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit