Learn More
Acetic acid, glacial, 99+%, Thermo Scientific Chemicals
Description
Good polar protic solvent, in its ester form it acts as a solvent for inks paints and coatings. Used in laboratory as a solvent for recrystallization, manufacture of camphor, cellulose acetate, polyvinyl acetate. It involves in Wagner-Meerwein rearrangement. Used for preparation of mono-lithium salt of 5-sulfoisophthalic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 16.6°C |
Density | 1.049 |
Boiling Point | 118.1°C |
Flash Point | 40°C (104°F) |
Assay Percent Range | 99+% |
Refractive Index | 1.3721 |
Quantity | 250 mL |
Solubility Information | Miscible with water,ethanol,acetone,toluene and immiscible with carbon disulphide. |
Formula Weight | 60.05 |
Physical Form | Liquid |
Show More |
Safety and Handling
Hazard Category | H226-H314 |
Precautionary Statement | P210-P233-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: II |
EINECSNumber | 200-580-7 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.