Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetic Acid, Glacial, ACS Reagent, Reagents
Click to view available options
Quantity:
500 mL
1 L
2.5 L
20 L
200 L
Specifications
Specifications
pH | <2 |
Boiling Point | 118°C |
Color | Colorless |
Grade | ACS |
Quantity | 20 L |
Packaging | HDPE Winpak |
Physical Form | Liquid |
Specific Gravity | 1.0496 |
Chemical Name or Material | Acetic Acid, Glacial, ACS Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction