Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid, Glacial, Aldehyde Free, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
2.5L
4L
Packaging:
Clear Glass Bottle
Clear Glass Bottle in Single Shipper
Clear PVC Coated Glass Bottle
Glass Bottle
Poly Bottle
PVC Coated Glass Bottle
64-19-7
C2H4O2
60.052
QTBSBXVTEAMEQO-UHFFFAOYSA-N
ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,glacial,acetasol,acide acetique,essigsaeure
176
CHEBI:15366
acetic acid
CC(=O)O
64-19-7
C2H4O2
60.052
QTBSBXVTEAMEQO-UHFFFAOYSA-N
ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,glacial,acetasol,acide acetique,essigsaeure
176
CHEBI:15366
acetic acid
CC(=O)O

High quality chemicals for laboratory and specialized industrial use

Formula Weight 60.05
Grade Meets ACS reagent chemical requirements
Packaging Poly Bottle
Specific Gravity 1L = 1.05kg
Quantity 4L
Flash Point 39.00°C
Chemical Name or Material Acetic Acid, Glacial, Aldehyde Free

Recommended Storage : IMPORTANT: Material will freeze if stored below 17°C (63°F)