Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid, Glacial, BAKER INSTRA-ANALYZED™ Plus, J.T.Baker™

Click to view available options
Quantity:
2.5L
4L
500mL
Packaging:
PFA Bottle
Poly Bottle

Chemical Identifiers

64-19-7
C2H4O2
60.052
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
CHEBI:15366

Acids for critical trace metal analysis; Quality is assured for up to 64 trace metals tested to very low parts-per-billion (ppb) levels

    For use in ICP (Inductively Coupled Plasma Spectrometry), Trace Metal Analysis; For Laboratory, Research, or Manufacturing Use
CAS 64-19-7
Molecular Formula C2H4O2
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
SMILES CC(=O)O
IUPAC Name acetic acid
Molecular Weight (g/mol) 60.052
PubChem CID 176
ChEBI CHEBI:15366
Formula Weight 60.05
Packaging PFA Bottle
Specific Gravity 1L = 1.04kg
Quantity 500mL
Flash Point 39.00°C
Chemical Name or Material Acetic Acid, Glacial
Grade BAKER INSTRA-ANALYZED™ Plus
Show More Show Less