Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid, Glacial, FCC, 99.5-100.5%, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
2.5 L
4 L
6 x 500 mL
20 L
4 x 2.5 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Poly-Coated Glass Bottle
64-19-7
C2H4O2
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
CC(O)=O
64-19-7
C2H4O2
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
CC(O)=O

A1009, 64-19-7, C2H4O2

Spectrum™ Acetic Acid, Glacial, FCC is used in the pickling of vegetables and other foods. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
UN Number UN2789
Percent Purity 99.5 to 100.5%
Lead (Pb) 0.5 mg/kg
Non_volatile Matter 0.005%
Packaging Drum
Quantity 200 L