Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid, Glacial, Multi-Compendial, U.S.P., J.T.Baker™

Click to view available options
Quantity:
500mL
2.5L
15L
Packaging:
Glass Bottle
Poly Pail
pvc coated glass bottle
64-19-7
C2H4O2
60.052
QTBSBXVTEAMEQO-UHFFFAOYSA-N
ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,glacial,acetasol,acide acetique,essigsaeure
176
CHEBI:15366
acetic acid
CC(=O)O
64-19-7
C2H4O2
60.052
QTBSBXVTEAMEQO-UHFFFAOYSA-N
ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,glacial,acetasol,acide acetique,essigsaeure
176
CHEBI:15366
acetic acid
CC(=O)O
Formula Weight 60.05
Grade Multi-Compendial/USP
Packaging pvc coated glass bottle
Specific Gravity 1L = 1.05kg
Quantity 2.5L
Flash Point 39.00°C
Chemical Name or Material Acetic Acid, Glacial