Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetic Acid, Glacial, Multi-Compendial, U.S.P., J.T. Baker™
Click to view available options
Quantity:
500 mL
2.5 L
15 L
Packaging:
Glass Bottle
Poly Pail
pvc coated glass bottle
Specifications
Formula Weight | 60.05 |
Grade | Multi-Compendial/USP |
Quantity | 500 mL |
Flash Point | 39.00°C |
Packaging | Glass Bottle |
Specific Gravity | 1L = 1.05kg |
Chemical Name or Material | Acetic Acid, Glacial |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction