Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid, Glacial (Optima™), Fisher Chemical™

Catalog Number A4651
Click to view available options
Quantity:
250 mL
500 mL
1 L
Packaging:
FEP Bottle
64-19-7
C2H4O2
60.05
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
176
CHEBI:15366
acetic acid
CC(O)=O
64-19-7
C2H4O2
60.05
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
176
CHEBI:15366
acetic acid
CC(O)=O
  • Certificate of Analysis included with each bottle.
  • Ideal for analysis by ICP-MS and high-resolution ICP-MS
  • Evaluated for 65 metals at low-level ppt concentrations.
  • Manufactured, purified, and packaged in a Class 1000 clean room.
  • Packaged in specially cleaned Teflon FEP for PFA bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
  • Three year shelf life from time of manufacture - available on COA.
TRUSTED_SUSTAINABILITY
pH 2.5
Melting Point 16°C
Boiling Point 117°C
Color Colorless
Linear Formula CH3COOH
UN Number 2789
Merck Index 15, 54
Formula Weight 60.05
Grade OPTIMA™
Density 1.048g/cm³
Quantity 1 L
Packaging FEP Bottle
Physical Form Liquid
Vapor Pressure 1.52 kPa at 20°C
Chemical Name or Material Acetic Acid
Show More Show Less
DOTInformation DOT Class 8, : Corrosive
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit