Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid, Glacial, Reagent, ACS, 99.7%, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
2.5 L
4 L
6 x 500 mL
20 L
4 x 2.5 L
4 x 4 L
6 x 2.5 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Bottle
Poly Drum
Poly-Coated Glass Bottle
64-19-7
C2H4O2
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
CC(O)=O
64-19-7
C2H4O2
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
CC(O)=O

A1010, 64-19-7, C2H4O2

Spectrum™ Acetic Acid, Glacial, Reagent, ACS , also known under the name ethanoic acid, it is an organic compound and a colorless liquid. It has a distinctive sour taste and pungent smell. It is the main component of Vinegar after water (8% by volume). Classified as a weak Acid, when concentrated it is corrosive and can attack the skin. In its Glacial form (undiluted), it is used as a polar protic solvent. As a solvent it is used for purification by recrystallization of organic compounds and in the production of terephtalic acid. As an ACS grade Reagent, Spectrum Chemical manufactured Acetic Acid is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and purity.
CAS Max % 100%
Chloride 1 ppm
UN Number UN2789
Percent Purity 99.7%
Heavy Metals (as Pb) 0.5 ppm
Packaging Poly Bottle
Residue after Evaporation 0.001%
Sulfate 1 ppm
Titratable Base 0.0004 meq/g
Quantity 6 x 500 mL