Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetic Acid, Glacial, Reagent, ACS, 99.7%, Spectrum™ Chemical
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
4 L
6 x 500 mL
20 L
4 x 2.5 L
4 x 4 L
6 x 2.5 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Bottle
Poly Drum
Poly-Coated Glass Bottle
Description
Spectrum™ Chemical Acetic Acid, Glacial, Reagent, ACS , also known under the name ethanoic acid, it is an organic compound and a colorless liquid. It has a distinctive sour taste and pungent smell. It is the main component of Vinegar after water (8% by volume). Classified as a weak Acid, when concentrated it is corrosive and can attack the skin. In its Glacial form (undiluted), it is used as a polar protic solvent. As a solvent it is used for purification by recrystallization of organic compounds and in the production of terephtalic acid. As an ACS grade Reagent, Spectrum Chemical manufactured Acetic Acid is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and purity.
Specifications
CAS Max % | 100% |
UN Number | UN2789 |
Percent Purity | 99.7% |
Grade | ACS |
Quantity | 4 x 4 L |
Packaging | Poly Bottle |
Residue after Evaporation | 0.001% |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction