Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid, Glacial, U.S.P. - F.C.C., J.T.Baker™

Click to view available options
Quantity:
500mL
2.5L
Packaging:
Glass Bottle
Glass Bottle, Single ship
pvc coated glass bottle
64-19-7
C2H4O2
60.052
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
CHEBI:15366
acetic acid
CC(=O)O
64-19-7
C2H4O2
60.052
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
CHEBI:15366
acetic acid
CC(=O)O
Formula Weight 60.05
Packaging Glass Bottle
Specific Gravity 1L = 1.05kg
Quantity 2.5L
Flash Point 39.00°C
Grade FCC/USP
Chemical Name or Material Acetic Acid, Glacial