Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic acid, potassium salt, 99+%, for biochemistry, ACROS Organics™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Plastic bottle
127-08-2
C2H3KO2
98.14
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium acetate,acetic acid, potassium salt,diuretic salt,potassium ethanoate,koac,octan draselny czech,potassiumacetate,acetic acid potassium salt,acok,fema no. 2920
517044
CHEBI:32029
potassium;acetate
CC(=O)[O-].[K+]
127-08-2
C2H3KO2
98.14
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium acetate,acetic acid, potassium salt,diuretic salt,potassium ethanoate,koac,octan draselny czech,potassiumacetate,acetic acid potassium salt,acok,fema no. 2920
517044
CHEBI:32029
potassium;acetate
CC(=O)[O-].[K+]
Sulfate 0.002% max
Phosphate 0.001% max
Chloride 0.003% max
Melting Point 292.0°C
Quantity 500g
Packaging Plastic bottle
Flash Point >250°C
Assay Percent Range 99+%
Linear Formula CH3CO2K
Merck Index 15,7726
Solubility Information (5 M in water) Clear colorless solution
Formula Weight 98.14
Percent Purity 99+%
Grade Biochemistry
Chemical Name or Material Acetic acid, potassium salt
Show More Show Less

EINECSNumber : 204-822-2