Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic acid, sodium salt, 99+%, ACS reagent, anhydrous, ACROS Organics™

Click to view available options
Quantity:
5g
500g
2.5kg
Packaging:
Glass bottle
Plastic Bottle
127-09-3
C2H3NaO2
82.034
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate,acetic acid, sodium salt,sodium acetate anhydrous,sodium acetate, anhydrous,acetic acid sodium salt,anhydrous sodium acetate,sodii acetas,sodium ethanoate,natrium aceticum,octan sodny czech
517045
CHEBI:32954
sodium;acetate
CC(=O)[O-].[Na+]
127-09-3
C2H3NaO2
82.034
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate,acetic acid, sodium salt,sodium acetate anhydrous,sodium acetate, anhydrous,acetic acid sodium salt,anhydrous sodium acetate,sodii acetas,sodium ethanoate,natrium aceticum,octan sodny czech
517045
CHEBI:32954
sodium;acetate
CC(=O)[O-].[Na+]
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Calcium (Ca) 50ppm max.
Linear Formula CH3CO2Na
Chlorine (Cl) 20ppm max.
Beilstein 02, 96
Fieser 01,1024; 05,591
Merck Index 15, 8709
Formula Weight 82.03
Physical Form Fine Crystalline Powder
Percent Purity ≥99%
Grade ACS Reagent
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Insoluble Matter 0.01% max.
Iron (Fe) 10ppm max.
Loss on Drying 1.0% max. (120°C)
Magnesium (Mg) 20ppm max.
Packaging Glass bottle
Phosphate 10ppm max.
Solubility Solubility in water: 500g/L (20°C). Other solubilities: slightly soluble in alcohol
Sulfate 30ppm max.
Melting Point 324.0°C
pH 7.0 to 9.2 (5% soln. at 25°C)
Color White
Quantity 5g
Flash Point >250°C
Chemical Name or Material Acetic acid, sodium salt
Show More Show Less

HYGROSCOPIC

EINECSNumber : 204-823-8

RTECSNumber : AJ4300010

TSCA : TSCA