Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic acid, sodium salt, 99+%, for analysis, anhydrous, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Plastic bottle
Plastic drum
127-09-3
C2H3NaO2
82.03
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
517045
CHEBI:32954
sodium;acetate
[Na+].CC([O-])=O
127-09-3
C2H3NaO2
82.03
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
517045
CHEBI:32954
sodium;acetate
[Na+].CC([O-])=O
Biological buffer
CAS Min % 99.0
CAS Max % 100.0
Aluminum (Al) 10ppm max.
Assay Percent Range 99+%
Calcium (Ca) 10ppm max.
Linear Formula CH3CO2Na
Chlorine (Cl) 10ppm max.
Formula Weight 82.03
Physical Form Crystals or Crystalline Powder
Percent Purity ≥99%
Grade Analysis
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 5ppm max.
Loss on Drying 1% max.
Magnesium (Mg) 5ppm max.
Packaging Plastic bottle
Phosphate 5ppm max.
Potassium (K) 200ppm max.
Solubility Solubility in water: 500g/L (20°C). Other solubilities: slightly soluble in alcohol
Sulfate 20ppm max.
Melting Point 324.0°C
pH 7.5 to 9.2 (5% soln. at 20°C)
Color White
Quantity 250g
Flash Point >250°C
Chemical Name or Material Acetic acid, sodium salt, p.a.
Show More Show Less

HYGROSCOPIC

EINECSNumber : 204-823-8

RTECSNumber : AJ4300010

TSCA : TSCA