Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic acid, sodium salt, 99%, for biochemistry, Anhydrous, Thermo Scientific Chemicals

Catalog Number AC212660010
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
127-09-3
C2H3NaO2
82.03
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
517045
CHEBI:32954
sodium acetate
[Na+].CC([O-])=O
127-09-3
C2H3NaO2
82.03
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
517045
CHEBI:32954
sodium acetate
[Na+].CC([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY
Absorbance (c= 0.1 M, water), (c= 0.1 M, water) at 0nm, 0.01 max. at 260nm, 0.01 max. at 280nm
Melting Point 324.0°C
Assay Percent Range 99%
Linear Formula CH3CO2Na
Solubility Information Solubility in water: 500g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 82.03
Percent Purity 99%
Grade Biochemical
Quantity 1 kg
Flash Point >250°C
Packaging Plastic bottle
Chemical Name or Material Acetic acid, sodium salt
Show More Show Less

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit