Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic acid, sodium salt, 99%, for biochemistry, Anhydrous, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
Packaging:
Plastic bottle
127-09-3
C2H3NaO2
82.034
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate,acetic acid, sodium salt,sodium acetate anhydrous,sodium acetate, anhydrous,acetic acid sodium salt,anhydrous sodium acetate,sodii acetas,sodium ethanoate,natrium aceticum,octan sodny czech
517045
CHEBI:32954
sodium;acetate
CC(=O)[O-].[Na+]
127-09-3
C2H3NaO2
82.034
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate,acetic acid, sodium salt,sodium acetate anhydrous,sodium acetate, anhydrous,acetic acid sodium salt,anhydrous sodium acetate,sodii acetas,sodium ethanoate,natrium aceticum,octan sodny czech
517045
CHEBI:32954
sodium;acetate
CC(=O)[O-].[Na+]
Biological buffer
Absorbance (c= 0.1 M, water),(c= 0.1 M, water) at 0nm,0.01 max. at 260nm,0.01 max. at 280nm
Assay Percent Range 99%
Linear Formula CH3CO2Na
Formula Weight 82.03
Percent Purity 99%
Grade Biochemical
Packaging Plastic bottle
Solubility Solubility in water: 500g/L (20°C). Other solubilities: slightly soluble in alcohol
Melting Point 324.0°C
Quantity 250g
Flash Point >250°C
Chemical Name or Material Acetic acid, sodium salt
Show More Show Less