Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic acid, sodium salt, 99+%, for mol biology, anhydrous, DNAse, RNAse and P, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
2.5kg
127-09-3
C2H3NaO2
82.034
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate,acetic acid, sodium salt,sodium acetate anhydrous,sodium acetate, anhydrous,acetic acid sodium salt,anhydrous sodium acetate,sodii acetas,sodium ethanoate,natrium aceticum,octan sodny czech
517045
CHEBI:32954
sodium;acetate
CC(=O)[O-].[Na+]
127-09-3
C2H3NaO2
82.034
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate,acetic acid, sodium salt,sodium acetate anhydrous,sodium acetate, anhydrous,acetic acid sodium salt,anhydrous sodium acetate,sodii acetas,sodium ethanoate,natrium aceticum,octan sodny czech
517045
CHEBI:32954
sodium;acetate
CC(=O)[O-].[Na+]
Biological buffer
Absorbance (0.1 M in water),(0.1 M in water) at 0nm,0.01 max. at 260nm,0.01 max. at 280nm
Absorbance Conditions (0.1M in water)
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula CH3CO2Na
Chlorine (Cl) 20ppm max.
DNase RNase Protease act none detected
Formula Weight 82.03
Physical Form Crystalline Solid
Percent Purity ≥99%
Grade Molecular Biology
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Insoluble in Water 0.01% max.
Iron (Fe) 10ppm max.
Loss on Drying 1% max. (120°C)
Packaging Plastic bottle
Phosphate 10ppm max.
Solubility Solubility in water: 500g/L (20°C). Other solubilities: slightly soluble in alcohol
Melting Point 324.0°C
pH 7 to 9.2 (5% soln. at 25°C)
Color White
Quantity 250g
Flash Point >250°C
Chemical Name or Material Acetic acid, sodium salt, Anhydrous, DNAse, RNAse and Protease Free
Show More Show Less