Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic acid, sodium salt trihydrate, 99+%, extra pure, ACROS Organics™

Click to view available options
Quantity:
1kg
5kg
Packaging:
Plastic Bottle
Plastic drum
127-09-3,6131-90-4
C2H3NaO2
82.034
MFCD00071557
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate,acetic acid, sodium salt,sodium acetate anhydrous,sodium acetate, anhydrous,acetic acid sodium salt,anhydrous sodium acetate,sodii acetas,sodium ethanoate,natrium aceticum,octan sodny czech
517045
CHEBI:32954
sodium;acetate
CC(=O)[O-].[Na+]
127-09-3,6131-90-4
C2H3NaO2
82.034
MFCD00071557
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate,acetic acid, sodium salt,sodium acetate anhydrous,sodium acetate, anhydrous,acetic acid sodium salt,anhydrous sodium acetate,sodii acetas,sodium ethanoate,natrium aceticum,octan sodny czech
517045
CHEBI:32954
sodium;acetate
CC(=O)[O-].[Na+]
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula CH3COONa·3H2O
Chlorine (Cl) 200ppm max.
Beilstein 02, 96
Fieser 01,1024; 05,591
Merck Index 15, 8709
Formula Weight 136.08
Physical Form Adhering Crystals
Percent Purity ≥99%
Grade Extra Pure
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Insoluble Matter 0.05% max.
Iron (Fe) 10ppm max.
Packaging Plastic drum
Potassium (K) 500ppm max.
Solubility Solubility in water: 762g/L (20 °C). Other solubilities: 1g/19mL alcohol
Sulfate 200ppm max.
Melting Point 58.0°C
Color White
Quantity 5kg
Chemical Name or Material Acetic acid, sodium salt trihydrate
Show More Show Less