Alfa Aesar™ Acetic acid-Trimethylpentane, 60/40 (v/v) soln.

Testing of peroxide

Manufacturer: Alfa Aesar™ 41895AP

Quantity 500mL

 View more versions of this product

Catalog No. AA41895AP

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00236632
    UN Number UN2924
    Chemical Name or Material Acetic acid-Trimethylpentane

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    MDL Number MFCD00236632
    UN Number UN2924
    Health Hazard 1 Danger
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H224-H304-H314-H318-H336
    Extremely flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause drowsiness or dizziness.
    Quantity 500mL
    Concentration 60/40 (v/v) soln.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Chemical Name or Material Acetic acid-Trimethylpentane

    GHS H Statement
    H224-H304-H314-H318-H336
    Extremely flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit