Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Anhydride 99+%, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
Packaging:
Glass bottle
108-24-7
C4H6O3
102.089
MFCD00008705
WFDIJRYMOXRFFG-UHFFFAOYSA-N
acetic anhydride,acetanhydride,acetic acid, anhydride,acetic oxide,acetyl oxide,ethanoic anhydride,acetyl ether,acetyl anhydride,acetic acid anhydride,anhydride acetique
7918
CHEBI:36610
acetyl acetate
CC(=O)OC(=O)C
108-24-7
C4H6O3
102.089
MFCD00008705
WFDIJRYMOXRFFG-UHFFFAOYSA-N
acetic anhydride,acetanhydride,acetic acid, anhydride,acetic oxide,acetyl oxide,ethanoic anhydride,acetyl ether,acetyl anhydride,acetic acid anhydride,anhydride acetique
7918
CHEBI:36610
acetyl acetate
CC(=O)OC(=O)C
Acylation reagent
Refractive Index 1.3890 to 1.3910 (20°C, 589nm)
Infrared Spectrum Authentic
Quantity 2.5L
Packaging Glass bottle
Color Colorless
Density 1.087g/mL
Assay Percent Range 99+%
Linear Formula (CH3CO)2O
Beilstein 02, 166
Fieser 01,3; 02,7; 05,3; 06,1; 07,1; 08,1; 09,1; 11,1; 12,1; 13,93; 14,150; 15,1
Merck Index 15, 55
Formula Weight 102.09
Physical Form Liquid
Percent Purity ≥99.0%
Chemical Name or Material Acetic anhydride, 99+%
Show More Show Less

GHS H Statement:
Fatal if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Corrosive to the respiratory tract.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-564-8

RTECSNumber : AK1925000

TSCA : TSCA