Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Anhydride, Reagent, ACS, 97%, Spectrum™

Click to view available options
Quantity:
125 mL
500 mL
2.5 L
4 L
6 x 500 mL
20 L
4 x 2.5 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Drum
108-24-7
C4H6O3
102.09
WFDIJRYMOXRFFG-UHFFFAOYSA-N
acetyl acetate
CC(=O)OC(C)=O
108-24-7
C4H6O3
102.09
WFDIJRYMOXRFFG-UHFFFAOYSA-N
acetyl acetate
CC(=O)OC(C)=O

A1015, 108-24-7, C4H6O3

Spectrum™ Acetic Anhydride, Reagent, ACS , or ethanoic anhydride as it is otherwise known is a colorless, strong smelling liquid chemical compound that is the simplest isolatable acid anhydride and commonly used as a reagent for organic synthesis. Mainly used for acetylations, its main use is for the conversion of cellulose to cellulose acetate and in the synthesis of aspirin. As an ACS grade Reagent, Spectrum Chemical manufactured acetic anhydride is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.
Residue after Evaporation 0.003%
Packaging Drum
Quantity 200 L
CAS Max % 100%
UN Number UN1715
Percent Purity 97%