Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Acetic hydrazide, 96%
Click to view available options
Quantity:
25g
100g
500g
Description
Applications
Acetic hydrazide is used as an antibiotic for the treatment of mycobacterium tuberculosis. It is also involved in the preparation of sunitinib b reacting with 5-fluoroisatin. Further, it inhibits mycolic acid biosynthesis. It serves as an important intermediate in organic synthesis such as nitric furosemide Long (nifuratrone) in medicine.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong bases.
Acetic hydrazide is used as an antibiotic for the treatment of mycobacterium tuberculosis. It is also involved in the preparation of sunitinib b reacting with 5-fluoroisatin. Further, it inhibits mycolic acid biosynthesis. It serves as an important intermediate in organic synthesis such as nitric furosemide Long (nifuratrone) in medicine.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong bases.
Specifications
Sensitivity | Hygroscopic |
Quantity | 25g |
Flash Point | >110°C (230°F) |
Melting Point | ~60°C to 68°C |
Boiling Point | 128°C to 129°C (18mmHg) |
Beilstein | 506165 |
Solubility Information | Soluble in water. |
Formula Weight | 74.08 |
Percent Purity | 96% |
Chemical Name or Material | Acetic hydrazide |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H228-H302+H332-H315-H319-H335
DOTInformation : Hazard Class: 4.1; Packaging Group: III
EINECSNumber : 213-948-7
RTECSNumber : AI1225000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen