Learn More
Acetic hydrazide, 96%, Thermo Scientific Chemicals
Description
Acetic hydrazide is used as an antibiotic for the treatment of mycobacterium tuberculosis. It is also involved in the preparation of sunitinib b reacting with 5-fluoroisatin. Further, it inhibits mycolic acid biosynthesis. It serves as an important intermediate in organic synthesis such as nitric furosemide Long (nifuratrone) in medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acetic hydrazide is used as an antibiotic for the treatment of mycobacterium tuberculosis. It is also involved in the preparation of sunitinib b reacting with 5-fluoroisatin. Further, it inhibits mycolic acid biosynthesis. It serves as an important intermediate in organic synthesis such as nitric furosemide Long (nifuratrone) in medicine.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong bases.
Specifications
Melting Point | ∼60°C to 68°C |
Boiling Point | 128°C to 129°C (18 mmHg) |
Flash Point | >110°C (230°F) |
Quantity | 25 g |
Beilstein | 506165 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 74.08 |
Percent Purity | 96% |
Chemical Name or Material | Acetic hydrazide |
Safety and Handling
Hazard Category | H228-H302+H332-H315-H319-H335 |
Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c |
DOTInformation | Hazard Class: 4.1; Packaging Group: III |
EINECSNumber | 213-948-7 |
RTECSNumber | AI1225000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.