Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetoacetyl coenzyme A sodium salt, >90%, MP Biomedicals™

Click to view available options
Quantity:
1mg
5mg
10mg
25mg
50mg
102029-52-7
C25H40N7NaO18P3S
874.597
RSOOKMDOLPTBRH-UHFFFAOYSA-N
133112705
S-[2-[3-[[4-[[[5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl]oxy-2-hydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyl] 3-oxobutanethioate;sodium
CC(=O)CC(=O)SCCNC(=O)CCNC(=O)C(C(C)(C)COP(=O)(O)OP(=O)(O)OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)O)O.[Na]
102029-52-7
C25H40N7NaO18P3S
874.597
RSOOKMDOLPTBRH-UHFFFAOYSA-N
133112705
S-[2-[3-[[4-[[[5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl]oxy-2-hydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyl] 3-oxobutanethioate;sodium
CC(=O)CC(=O)SCCNC(=O)CCNC(=O)C(C(C)(C)COP(=O)(O)OP(=O)(O)OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)O)O.[Na]
Quantity 5mg
Color Yellow
Assay Percent Range >90%
Formula Weight 901.6 (free acid)
Physical Form Solid
Chemical Name or Material Acetoacetyl coenzyme A

Recommended Storage : Store at -20°C.