Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, 99.5+%, for analysis, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
500mL
1L
2.5L
5L
25L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Metal Drum
Plastic Bottle
Plastic drum
67-64-1
C3H6O
58.08
MFCD00008765
CSCPPACGZOOCGX-UHFFFAOYSA-N
acetone,2-propanone,propanone,dimethyl ketone,methyl ketone,dimethylformaldehyde,pyroacetic ether,beta-ketopropane,dimethylketal,chevron acetone
180
CHEBI:15347
propan-2-one
CC(=O)C
67-64-1
C3H6O
58.08
MFCD00008765
CSCPPACGZOOCGX-UHFFFAOYSA-N
acetone,2-propanone,propanone,dimethyl ketone,methyl ketone,dimethylformaldehyde,pyroacetic ether,beta-ketopropane,dimethylketal,chevron acetone
180
CHEBI:15347
propan-2-one
CC(=O)C
For analysis

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

CAS Min % 99.5
CAS Max % 100.0
Additional Information Vapor Pressure: 247mbar at 20°C
Acidity Alkalinity 0.0001 meq/g max.
Aldehyde 0.002% max.
Assay Percent Range 99.5+%
Linear Formula CH3COCH3
Beilstein 01, 635
Fieser 02,13; 03,4; 06,9
Merck Index 15, 65
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol, dimethylformamide,, chloroform, ether and most oils
Cadmium (Cd) 0.05ppm max.
Formula Weight 58.08
Physical Form Liquid
Percent Purity ≥99.5%
Grade Analysis
Copper (Cu) 0.1ppm max.
Density 0.7900g/mL
Iron (Fe) 0.5ppm max.
Lead (Pb) 0.1ppm max.
Magnesium (Mg) 0.1ppm max.
Packaging Metal Drum
Refractive Index 1.3585
Residue after Evaporation 0.001% max.
Specific Gravity 0.79
Substances Reducing Permanganate 5ppm max. (O)
Trace Analysis 0.05ppm max.
Water 0.5% max. (Coulometric)
Flash Point −20°C
Viscosity 0.32 mPa.s (20°C)
Decomposition Information >4°C
Boiling Point 56.0°C
Color Undesignated
Melting Point -95.0°C
Quantity 25L
Chemical Name or Material Acetone, For analysis
Show More Show Less

GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger