Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, 99.5%, MilliporeSigma™

Click to view available options
Quantity:
1 L
20 L
Packaging:
Glass Bottle, Pre-Packed
Plastic Drum
67-64-1
58.08 g/mol
67-64-1
58.08 g/mol
Assay Percent Range 99.5% min
Packaging Glass Bottle, Pre-Packed
Residue after Evaporation 5 ppm max
Vapor Pressure 233 hPa (20°C)
Flash Point < −20°C
Boiling Point 56.2°C (1013 hPa)
Melting Point −95.4°C
pH 5 to 6 (395 g/L, H2O, 20°C)
Quantity 1 L

Recommended Storage : Store at room temperature