Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, ACS reagent, ≥99.5%, Honeywell Riedel-de Haën™

Click to view available options
Quantity:
500mL
1L
2.5L
20L
4 x 2.5L
6 x 1L
Packaging:
Glass Bottle
Stainless Steel Can
67-64-1
C3H6O
58.08
MFCD00008765
CSCPPACGZOOCGX-UHFFFAOYSA-N
acetone,2-propanone,propanone,dimethyl ketone,methyl ketone,dimethylformaldehyde,pyroacetic ether,beta-ketopropane,dimethylketal,chevron acetone
180
CHEBI:15347
propan-2-one
CC(=O)C
67-64-1
C3H6O
58.08
MFCD00008765
CSCPPACGZOOCGX-UHFFFAOYSA-N
acetone,2-propanone,propanone,dimethyl ketone,methyl ketone,dimethylformaldehyde,pyroacetic ether,beta-ketopropane,dimethylketal,chevron acetone
180
CHEBI:15347
propan-2-one
CC(=O)C

ACS reagent,≥99.5%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Linear Formula CH3COCH3
UN Number UN1090
Beilstein 635680
Formula Weight 58.08
Percent Purity >99.5%
Grade ACS Reagent
Density 0.791 g/mL at 25 °C(lit.)
Packaging Glass Bottle
Flash Point −17°C
Boiling Point 55°C to 57°C
Melting Point -94°C (literature)
Quantity 500mL
Chemical Name or Material Acetone, ACS reagent, ≥99.5%
Assay acc. to ACS
Show More Show Less

ShelfLife : 1080 days from date of manufacture

DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetone

EINECSNumber : 200-662-2

Recommended Storage : Room Temp