Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetone-Alcohol, 1:1, Decolorizer, Spectrum™ Chemical
Specifications
CAS | 67-64-1, 64-17-5, 67-63-0 |
---|---|
Packaging | Poly Bottle |
Grade | Chemical Solution |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
18-608-407
|
Spectrum Chemical Manufacturing Corporation
A096500ML |
500 mL | Poly Bottle | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
18-608-405
|
Spectrum Chemical Manufacturing Corporation
A0961LT |
1 L | Poly Bottle | Item Discontinued Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier. | N/A | ||||
18-608-406
|
Spectrum Chemical Manufacturing Corporation
A0964LT |
4 L | Poly Bottle | Item Discontinued Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier. | N/A | ||||
Specifications
67-64-1, 64-17-5, 67-63-0 | |
Poly Bottle | |
Chemical Solution |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title