Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetone, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.9% (excluding water), Honeywell Burdick & Jackson
Click to view available options
Quantity:
20 L
Packaging:
Returnable NOWPAK™ II
Specifications
CAS Max % | 100.0000% |
Density | 0.79 g/cm3 |
Boiling Point | 56°C |
Quantity | 20 L |
Flash Point | −20°C |
Packaging | Returnable NOWPAK™ II |
Beilstein | 635680 |
Formula Weight | 58.08 |
Percent Purity | >99.9% |
Grade | HPLC |
Show More |
Safety and Handling
ShelfLife | 547 days from date of manufacture |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction