Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
500mL
Packaging:
Aluminum Bottle
HDPE Pressure Bottom Poly Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC- Coated Amber Glass Bottle

Chemical Identifiers

67-64-1
C3H6O
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
180
CHEBI:15347

High quality chemicals for laboratory and specialized industrial use

    For Laboratory,Research,or Manufacturing Use
CAS 67-64-1
Molecular Formula C3H6O
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES CC(=O)C
IUPAC Name propan-2-one
Molecular Weight (g/mol) 58.08
PubChem CID 180
ChEBI CHEBI:15347
Formula Weight 58.08
Packaging Aluminum Bottle
Specific Gravity 1L = 0.79kg
Flash Point -20°C (-4°C)
Boiling Point ≤56.1°C
Quantity 4L
Chemical Name or Material Acetone
Grade Baker Analyzed™ ACS Reagent, Meets ACS reagent chemical requirements
Show More Show Less